Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011132.2 | NP_035262.2 | 21 | 2404 | |
GenBank mRNA | AF123502.1 | - | 21 | 2378 | |
AK141674.1 | - | 21 | 2365 | ||
BC048166.1 | - | 21 | 2339 | 6410453 | |
BC063246.1 | - | 21 | 2339 | 6410453 |