Gene Transcripts

Gene Symbol
Prl
Entrez Gene ID
19109
Gene Name
prolactin
Gene Aliases
AV290867, Prl1a1
Chromosome Location
Chr. 13: 27057570 - 27065204 on Build GRCm38
UniGene ID
Mm.1270
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163530.1 NP_001157002.1 2 240
NM_011164.2 NP_035294.2 2 243
GenBank mRNA AK017520.1 - 2 236
AK017521.1 - 2 237
AK017547.1 - 2 241
AK017562.1 - 2 228
AK017579.1 - 2 237
AK017584.1 - 2 234
AK077493.1 - 2 240
AK133645.1 - 2 234
AK133736.1 - 2 237
AK133752.1 - 2 237
AK161553.1 - 2 233
AK161557.1 - 2 237
AK161565.1 - 2 237
AK161594.1 - 2 234
AK161619.1 - 2 241
BC061141.1 - 2 236 30250937
X02892.1 - 2 235
X04418.1 - 2 237

View Full Product Details