Gene Transcripts
- Gene Symbol
- Ptp4a3
- Entrez Gene ID
- 19245
- Gene Name
- protein tyrosine phosphatase 4a3
- Gene Aliases
- AV088979, Prl-3, pPtp4a3
- Chromosome Location
- Chr. 15: 73723145 - 73758766 on Build GRCm38
- UniGene ID
- Mm.390807
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001166388.1 |
NP_001159860.1
|
3, 4 |
1630 |
|
NM_001166389.1 |
NP_001159861.1
|
3, 4 |
1361 |
|
NM_001166390.1 |
NP_001159862.1
|
2, 3 |
462 |
|
NM_008975.3 |
NP_033001.2
|
3, 4 |
1430 |
|
GenBank mRNA |
AF035645.1 |
- |
2, 3 |
411 |
|
AK003954.1 |
- |
2, 3 |
168 |
|
AK004562.1 |
- |
3, 4 |
1040 |
|
AK014601.1 |
- |
3, 4 |
1044 |
|
AK143702.1 |
- |
3, 4 |
1082 |
|
AK147489.1 |
- |
1, 2 |
5823 |
|
AK172192.1 |
- |
3, 4 |
1084 |
|
BC027445.1 |
- |
2, 3 |
241 |
4482106 |
BC066043.1 |
- |
3, 4 |
1630 |
6415021 |
CK621052.1 |
- |
2, 3 |
462 |
|
CT010215.1 |
- |
2, 3 |
196 |
|
View Full Product Details