Gene Transcripts

Gene Symbol
Abcd4
Entrez Gene ID
19300
Gene Name
ATP-binding cassette, sub-family D (ALD), member 4
Gene Aliases
P69r, P70R, Pxmp1l
Chromosome Location
Chr. 12: 84602531 - 84617479 on Build GRCm38
UniGene ID
Mm.229109
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008992.2 NP_033018.2 12, 13 1484
GenBank mRNA AJ001166.1 - 12, 13 1388
AK028420.1 - 13 1541
BC050102.1 - 12, 13 1592 30012556

View Full Product Details