Gene Transcripts

Gene Symbol
Rab1
Entrez Gene ID
19324
Gene Name
RAB1, member RAS oncogene family
Gene Aliases
Gtbp, Rab-1, Rab1A, Ypt1, mKIAA3012
Chromosome Location
Chr. 11: 20201602 - 20226856 on Build GRCm38
UniGene ID
Mm.271944
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008996.3 NP_033022.1 3 187
GenBank mRNA AB232583.1 - 3 159
AF226873.1 - 3 187
AK034408.1 - 3 355
AK050526.1 - 3 351
AK129477.1 - 3 345
AK151085.1 - 3 351
AK152978.1 - 3 356
AK159425.1 - 3 351
AK166249.1 - 3 348
AK167150.1 - 3 355
AK197899.1 - 246
BC002077.1 - 3 354 3592802
Y00094.1 - 3 288

View Full Product Details