Gene Transcripts

Gene Symbol
Rag1
Entrez Gene ID
19373
Gene Name
recombination activating gene 1
Gene Aliases
Rag-1
Chromosome Location
Chr. 2: 101638252 - 101649532 on Build GRCm38
UniGene ID
Mm.828
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009019.2 NP_033045.2 2 1715
GenBank mRNA BC138342.1 - 2 1640
M29475.1 - 2 1684

View Full Product Details