Gene Transcripts

Gene Symbol
Rrh
Entrez Gene ID
20132
Gene Name
retinal pigment epithelium derived rhodopsin homolog
Gene Aliases
Chromosome Location
Chr. 3: 129808575 - 129822505 on Build GRCm38
UniGene ID
Mm.278758
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009102.3 NP_033128.1 3, 4 430
GenBank mRNA AF012271.1 - 3, 4 434
AK021379.1 - 1, 2 140
AK044717.1 - 418
BC046288.1 - 4, 5 556 6468540

View Full Product Details