Gene Transcripts

Gene Symbol
Six4
Entrez Gene ID
20474
Gene Name
sine oculis-related homeobox 4
Gene Aliases
AI047561, AREC3, TrexBF
Chromosome Location
Chr. 12: 73099609 - 73114774 on Build GRCm38
UniGene ID
Mm.249575
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011382.2 NP_035512.1 2 1119
GenBank mRNA BC137931.1 - 2 1102
BC137934.1 - 2 1102 8860953
D50416.1 - 2 1119
D50417.1 - 4 1034
D50418.1 - 4 892

View Full Product Details