Gene Transcripts

Gene Symbol
Sox6
Entrez Gene ID
20679
Gene Name
SRY (sex determining region Y)-box 6
Gene Aliases
AI987981, SOX-LZ
Chromosome Location
Chr. 7: 115470872 - 116040484 on Build GRCm38
UniGene ID
Mm.323365
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025559.3 NP_001020730.1 13 1681
NM_001025560.2 NP_001020731.1 13 1795
NM_001277326.1 NP_001264255.1 13 1776
NM_001277327.1 NP_001264256.1 12 1836
NM_001277328.1 NP_001264257.1 11 1541
NM_011445.4 NP_035575.1 14 1829
GenBank mRNA AJ010605.1 - 2 108
AK029696.1 - 11 1469
AK030713.1 - 13 1796
AK084290.1 - 13 1837
AK132409.1 - 13 1742
BC067407.1 - 12 1833 30094413
D61689.1 - 11 1537
U32614.1 - 14 1833

View Full Product Details