Gene Transcripts

Gene Symbol
Spint1
Entrez Gene ID
20732
Gene Name
serine protease inhibitor, Kunitz type 1
Gene Aliases
HAI-1
Chromosome Location
Chr. 2: 119237360 - 119249535 on Build GRCm38
UniGene ID
Mm.104955
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016907.3 NP_058603.2 11 1531
GenBank mRNA AF099018.1 - 11 1486
AK017342.1 - 10 1413
BC005769.1 - 10 1418 2650110

View Full Product Details