Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025287.2 | NP_079563.2 | 7 | 712 | |
GenBank mRNA | AB071989.1 | - | 5 | 521 | |
AK004669.1 | - | 7 | 682 | ||
AK028201.1 | - | 6 | 848 | ||
AK030746.1 | - | 6 | 899 | ||
AK132383.1 | - | 6 | 888 | ||
AK159482.1 | - | 6 | 865 | ||
AK160858.1 | - | 6 | 810 | ||
AK166581.1 | - | 6 | 822 | ||
AK168546.1 | - | 6 | 901 | ||
AK189454.1 | - | 2 | 113 | ||
AY538613.1 | - | 5 | 524 | ||
BC043131.1 | - | 6 | 731 | 6811159 | |
BC045205.1 | - | 7 | 682 | 4988521 | |
BC064809.1 | - | 6 | 836 |