Gene Transcripts

Gene Symbol
Exog
Entrez Gene ID
208194
Gene Name
endo/exonuclease (5'-3'), endonuclease G-like
Gene Aliases
AW557704, ENGL-B, ENGL-a, Endogl1, Endogl2, Engl, Engla, Englb
Chromosome Location
Chr. 9: 119444923 - 119465518 on Build GRCm38
UniGene ID
Mm.101578
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172136.1 NP_001165607.1 7 1006
NM_172456.3 NP_766044.1 6 971
GenBank mRNA AK028887.1 - 6 935
AK034654.1 - 7 968
AK051300.1 - 7 970
AK148426.1 - 6 952
AK162566.1 - 6 934
AK163555.1 - 6 935
AK184683.1 - 1 179
BC052171.1 - 5 777
BC059920.1 - 7 949
BC064730.1 - 7 1021
BC094589.1 - 4 3457
BC119024.1 - 6 935 8733773
BC144729.1 - 7 970

View Full Product Details