Gene Transcripts

Gene Symbol
Eif2b1
Entrez Gene ID
209354
Gene Name
eukaryotic translation initiation factor 2B, subunit 1 (alpha)
Gene Aliases
26kDa, D5Ertd406e, EIF2B, EIF2BA
Chromosome Location
Chr. 5: 124570214 - 124579057 on Build GRCm38
UniGene ID
Mm.28839
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145371.3 NP_663346.1 4 368
GenBank mRNA AK088066.1 - 4 367
AK133695.1 - 4 369
BC003426.1 - 4 368 2615801

View Full Product Details