Gene Transcripts

Gene Symbol
Plekhg6
Entrez Gene ID
213522
Gene Name
pleckstrin homology domain containing, family G (with RhoGef domain) member 6
Gene Aliases
BC026778, Gm155
Chromosome Location
Chr. 6: 125362651 - 125380951 on Build GRCm38
UniGene ID
Mm.333396
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198604.2 NP_941006.2 6 969
GenBank mRNA AK153619.1 - 6 1259
BC026778.1 - 6 972 5133546

View Full Product Details