Gene Transcripts

Gene Symbol
Atg9b
Entrez Gene ID
213948
Gene Name
autophagy related 9B
Gene Aliases
Apg912, Apg9l2, Apgdc2, Gm574, Nos3as, eONE, sONE
Chromosome Location
Chr. 5: 24384181 - 24392143 on Build GRCm38
UniGene ID
Mm.332252
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002897.3 NP_001002897.2 9 2402
GenBank mRNA AY495541.1 - 9 2402
AY515312.1 - 4 575
AY515313.1 - 9 2402

View Full Product Details