Gene Transcripts

Gene Symbol
Slc24a1
Entrez Gene ID
214111
Gene Name
solute carrier family 24 (sodium/potassium/calcium exchanger), member 1
Gene Aliases
Chromosome Location
Chr. 9: 64922861 - 64951607 on Build GRCm38
UniGene ID
Mm.156124
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144813.1 NP_659062.1 2 428
GenBank mRNA BC016094.1 - 2 428

View Full Product Details