Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153139.4 | NP_694779.3 | 9, 10 | 1144 | |
GenBank mRNA | AF453743.1 | - | 8, 9 | 1130 | |
AY211262.1 | - | 9, 10 | 1114 | ||
BC138556.1 | - | 9, 10 | 1108 | 8861576 | |
BC138558.1 | - | 9, 10 | 1108 | 8861578 |