Gene Transcripts

Gene Symbol
Appl2
Entrez Gene ID
216190
Gene Name
adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 2
Gene Aliases
Dip3b
Chromosome Location
Chr. 10: 83600033 - 83648854 on Build GRCm38
UniGene ID
Mm.282985
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145220.2 NP_660255.1 9 759
GenBank mRNA AK154525.1 - 9 783
AK160075.1 - 9 738
AK160105.1 - 9 735
AK161003.1 - 9 766
AK168403.1 - 9 718
AK168748.1 - 9 715
AK171512.1 - 9 715
AY113706.1 - 9 761
BC048906.1 - 9 697 6475576

View Full Product Details