Gene Transcripts

Gene Symbol
Cntrob
Entrez Gene ID
216846
Gene Name
centrobin, centrosomal BRCA2 interacting protein
Gene Aliases
9830165K03Rik, Lip8
Chromosome Location
Chr. 11: 69299491 - 69323893 on Build GRCm38
UniGene ID
Mm.28774
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172560.3 NP_766148.2 12 2598
GenBank mRNA AK036710.1 - 11 2418
AK147774.1 - 12 2488
BC058512.1 - 12 1431 6817014

View Full Product Details