Gene Transcripts

Gene Symbol
Pomt2
Entrez Gene ID
217734
Gene Name
protein-O-mannosyltransferase 2
Gene Aliases
A830009D15Rik, AW046274
Chromosome Location
Chr. 12: 87106861 - 87147902 on Build GRCm38
UniGene ID
Mm.388249
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153415.3 NP_700464.2 16 2192
GenBank mRNA AF246235.1 - 16 1796
AK043573.1 - 16 1893
AK043713.1 - 16 1734
AK050915.1 - 16 1761
AK076813.1 - 16 2195
AY090481.1 - 16 2192
AY090482.1 - 16 1792
BC052045.1 - 16 1764 6402328
BC062965.1 - 16 1764
BC079589.1 - 16 1762

View Full Product Details