Gene Transcripts

Gene Symbol
Traf2
Entrez Gene ID
22030
Gene Name
TNF receptor-associated factor 2
Gene Aliases
AI325259
Chromosome Location
Chr. 2: 25517982 - 25546940 on Build GRCm38
UniGene ID
Mm.3399
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290413.1 NP_001277342.1 9 1208
NM_009422.3 NP_033448.2 9 1187
GenBank mRNA AF027570.1 - 8 1099
AK038136.1 - 9 1136
AK052934.1 - 9 1136
AK151653.1 - 9 1125
AK152174.1 - 9 1155
AK159581.1 - 9 1157
AK159585.1 - 1134
BC003801.1 - 8 1113 3492839
BC060625.1 - 7 3144
L35303.1 - 9 1124

View Full Product Details