Gene Transcripts

Gene Symbol
Trhr
Entrez Gene ID
22045
Gene Name
thyrotropin releasing hormone receptor
Gene Aliases
TRH-R1
Chromosome Location
Chr. 15: 44195924 - 44235912 on Build GRCm38
UniGene ID
Mm.309350
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013696.2 NP_038724.1 2 1104
GenBank mRNA AK134591.1 - 2 1105
BC109005.1 - 2 989 40048176
BC109006.1 - 2 989 40048178
BC128269.1 - 2 989 40048433
M59811.1 - 2 1013
M94384.1 - 2 1013

View Full Product Details