Gene Transcripts

Gene Symbol
Vdac3
Entrez Gene ID
22335
Gene Name
voltage-dependent anion channel 3
Gene Aliases
Chromosome Location
Chr. 8: 22577075 - 22593813 on Build GRCm38
UniGene ID
Mm.227704
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198998.1 NP_001185927.1 7 489
NM_011696.2 NP_035826.1 6 486
GenBank mRNA AK002296.1 - 6 434
AK083785.1 - 7 417
AK146957.1 - 7 442
AK150741.1 - 6 428
AK150828.1 - 7 417
AK159431.1 - 6 428
AK161283.1 - 6 415
AK167781.1 - 6 431
AK168815.1 - 7 434
AK169099.1 - 7 442
AK169228.1 - 6 414
BC004743.1 - 6 384 3583800
BC089336.1 - 7 406 30357677
U30839.1 - 6 431

View Full Product Details