Gene Transcripts

Gene Symbol
Rbm11
Entrez Gene ID
224344
Gene Name
RNA binding motif protein 11
Gene Aliases
A330018F01
Chromosome Location
Chr. 16: 75592844 - 75602825 on Build GRCm38
UniGene ID
Mm.273214
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198302.2 NP_938044.1 5 652
GenBank mRNA AK039297.1 - 5 594
BC050779.1 - 5 699 6771154

View Full Product Details