Gene Transcripts

Gene Symbol
Adamts10
Entrez Gene ID
224697
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 10
Gene Aliases
9430006O18, AW045948, Adam-ts10, Adamts-10, Znmp
Chromosome Location
Chr. 17: 33524176 - 33553782 on Build GRCm38
UniGene ID
Mm.29304
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172619.3 NP_766207.2 4, 5 755
GenBank mRNA AK034565.1 - 4, 5 730
AK042525.1 - 4, 5 743
AK076334.1 - 4, 5 729
BC056427.1 - 4, 5 725
BC082773.1 - 4, 5 747
BC100558.1 - 3, 4 509
BC145353.1 - 3, 4 662
BC145354.1 - 3, 4 662
BC145355.1 - 3, 4 662

View Full Product Details