Gene Transcripts

Gene Symbol
Kdm2a
Entrez Gene ID
225876
Gene Name
lysine (K)-specific demethylase 2A
Gene Aliases
100043628, 5530401A10Rik, AA589516, AW536790, Cxxc8, Fbl11, Fbl7, Fbxl11, Gm4560, Jhdm1, Jhdm1a, lalina
Chromosome Location
Chr. 19: 4316144 - 4398280 on Build GRCm38
UniGene ID
Mm.31941
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001984.2 NP_001001984.2 12, 13 2295
GenBank mRNA AK173084.1 - 1, 2 113
BC037117.1 - 5, 6 844
BC076576.1 - 12, 13 1861 6840591

View Full Product Details