Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011731.3 | NP_035861.2 | 11 | 2088 | |
GenBank mRNA | AF075261.1 | - | 11 | 1912 | |
AK076806.1 | - | 12 | 1819 | ||
AK143893.1 | - | 11 | 2061 | ||
AK144038.1 | - | 11 | 2089 | ||
AK163340.1 | - | 12 | 1977 | ||
AK164706.1 | - | 11 | 1933 | ||
AK164713.1 | - | 9 | 3965 | ||
BC013484.1 | - | 11 | 1999 | 4242168 |