Gene Transcripts

Gene Symbol
Smg7
Entrez Gene ID
226517
Gene Name
Smg-7 homolog, nonsense mediated mRNA decay factor (C. elegans)
Gene Aliases
9430023P16Rik, mKIAA0250
Chromosome Location
Chr. 1: 152836995 - 152902686 on Build GRCm38
UniGene ID
Mm.270775
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005507.2 NP_001005507.1 17 2606
NM_001160256.1 NP_001153728.1 17 2468
NM_001160257.1 NP_001153729.1 17 2458
GenBank mRNA AK129101.1 - 17 2458
AK145908.1 - 2 347
BC082789.1 - 17 2593 30105222
BC086651.1 - 17 2468 6851591

View Full Product Details