Gene Transcripts

Gene Symbol
Plekhb2
Entrez Gene ID
226971
Gene Name
pleckstrin homology domain containing, family B (evectins) member 2
Gene Aliases
2310009M15Rik, AU018239, Evt2, Phdc, evt-2
Chromosome Location
Chr. 1: 34849959 - 34879585 on Build GRCm38
UniGene ID
Mm.292751
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145516.2 NP_663491.1 4 374
NM_175421.1 NP_780630.1 4 374
GenBank mRNA AF189817.1 - 4 334
AK039771.1 - 4 342
AK082118.1 - 4 327
AK148653.1 - 4 357
AK149868.1 - 4 334
AK150074.1 - 4 334
AK166091.1 - 4 342
BC057994.1 - 4 342 5344583
CK625615.1 - 4 374

View Full Product Details