Gene Transcripts

Gene Symbol
Ncoa5
Entrez Gene ID
228869
Gene Name
nuclear receptor coactivator 5
Gene Aliases
CIA
Chromosome Location
Chr. 2: 165000357 - 165034868 on Build GRCm38
UniGene ID
Mm.233080
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144892.1 NP_659141.1 5, 6 748
GenBank mRNA AK049242.1 - 2, 3 152
AK173226.1 - 3, 4 582
BC017152.1 - 5, 6 748 4511696

View Full Product Details