Gene Transcripts

Gene Symbol
Haus6
Entrez Gene ID
230376
Gene Name
HAUS augmin-like complex, subunit 6
Gene Aliases
6230416J20Rik, 9830144E06, D4Ertd27e, mKIAA1574
Chromosome Location
Chr. 4: 86578855 - 86612076 on Build GRCm38
UniGene ID
Mm.74730
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173400.2 NP_775576.2 8 969
GenBank mRNA AK051016.1 - 8 990
AK077681.1 - 8 991
AK169222.1 - 8 971
AK173206.1 - 8 941
BC064725.1 - 3 166
BC068237.1 - 8 969 30550558

View Full Product Details