Gene Transcripts

Gene Symbol
Lrrc41
Entrez Gene ID
230654
Gene Name
leucine rich repeat containing 41
Gene Aliases
AA409966, AW555107, D630045E04Rik, D730026A16Rik, MUF1
Chromosome Location
Chr. 4: 116075465 - 116097043 on Build GRCm38
UniGene ID
Mm.260786
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK028000.1 - 5 615
AK052762.1 - 8 1918
AK154613.1 - 8 2064
BC005765.1 - 7 1785
BC007169.1 - 5 943
BC022976.1 - 8 1996 5362925
BC024940.1 - 8 1993
BC071258.1 - 8 1929

View Full Product Details