Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172701.2 | NP_766289.2 | 1, 2 | 231 | |
GenBank mRNA | AB154826.1 | - | 1, 2 | 184 | |
AK077467.1 | - | 1, 2 | 197 | ||
AK077909.1 | - | 1, 2 | 152 | ||
BC045150.1 | - | 1, 2 | 177 | 6309923 |