Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145958.1 | NP_001139430.1 | 32 | 5143 | |
NM_172122.2 | NP_742120.2 | 33 | 5396 | ||
GenBank mRNA | AF527975.1 | - | 33 | 5396 | |
AK032335.1 | - | 4 | 1061 | ||
AK122285.1 | - | 24 | 4639 | ||
BC027090.1 | - | 6 | 937 | ||
BC034536.1 | - | 10 | 2301 | ||
BC036729.1 | - | 18 | 3038 | ||
BC038074.1 | - | 2 | 278 | ||
BC054054.1 | - | 31 | 5124 | 6394697 |