Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172711.2 | NP_766299.1 | 8 | 981 | |
GenBank mRNA | AK032597.1 | - | 7 | 1989 | |
AK047411.1 | - | 8 | 850 | ||
AK053420.1 | - | 8 | 955 | ||
AK084627.1 | - | 8 | 962 | ||
AK140534.1 | - | 8 | 940 | ||
AK160956.1 | - | 8 | 1000 | ||
AK165012.1 | - | 8 | 970 | ||
BC039660.1 | - | 1 | 65 | ||
BC057962.1 | - | 8 | 935 | 5353891 |