Gene Transcripts

Gene Symbol
Mat2a
Entrez Gene ID
232087
Gene Name
methionine adenosyltransferase II, alpha
Gene Aliases
D630045P18Rik
Chromosome Location
Chr. 6: 72432796 - 72439608 on Build GRCm38
UniGene ID
Mm.29815
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145569.4 NP_663544.1 6 858
GenBank mRNA AB070266.1 - 6 822
AK034757.1 - 6 859
AK052764.1 - 6 857
AK079420.1 - 6 864
AK133960.1 - 6 857
AK144189.1 - 6 861
AK145407.1 - 6 857
AK150040.1 - 6 857
AK151281.1 - 6 857
AK160869.1 - 6 857
AK161852.1 - 6 857
AK164137.1 - 6 857
AK166671.1 - 6 857
AK168155.1 - 6 857
AK169705.1 - 6 857
BC003451.1 - 6 798 2655444
BC058360.1 - 6 839 6821781

View Full Product Details