Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173789.4 | NP_776150.2 | 4 | 978 | |
GenBank mRNA | AB098077.1 | - | 4 | 429 | |
AB120968.1 | - | 4 | 978 | ||
AK044070.1 | - | 3 | 529 | ||
BC103599.1 | - | 4 | 863 | 40045925 | |
BC103600.1 | - | 4 | 863 | 40045926 | |
BC103601.1 | - | 4 | 863 | 40045923 | |
DQ294234.1 | - | 4 | 441 |