Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145614.4 | NP_663589.3 | 13 | 2006 | |
GenBank mRNA | AK032124.1 | - | 13 | 2007 | |
AK043771.1 | - | 13 | 1695 | ||
AK166090.1 | - | 13 | 1777 | ||
AY044265.1 | - | 13 | 1443 | ||
BC003202.1 | - | 2 | 134 | ||
BC026680.1 | - | 13 | 1754 | 5059786 | |
BC031495.1 | - | 13 | 1727 | 2651095 | |
BC069862.1 | - | 13 | 1749 | 5388446 |