Gene Transcripts

Gene Symbol
Klhl4
Entrez Gene ID
237010
Gene Name
kelch-like 4
Gene Aliases
B130014A11, C130018J01Rik
Chromosome Location
Chr. X: 114474333 - 114561129 on Build GRCm38
UniGene ID
Mm.132583
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172781.2 NP_766369.2 6 1397
GenBank mRNA AK081937.1 - 6 1210
BC057137.1 - 6 1128
BC099954.1 - 6 1224 30081872

View Full Product Details