Gene Transcripts
- Gene Symbol
- Klhl4
- Entrez Gene ID
- 237010
- Gene Name
- kelch-like 4
- Gene Aliases
- B130014A11, C130018J01Rik
- Chromosome Location
- Chr. X: 114474333 - 114561129 on Build GRCm38
- UniGene ID
- Mm.132583
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290477.1 |
NP_001277406.1
|
4, 5 |
1018 |
|
NM_001290478.1 |
NP_001277407.1
|
4, 5 |
1018 |
|
NM_172781.2 |
NP_766369.2
|
4, 5 |
1018 |
|
GenBank mRNA |
AK044930.1 |
- |
4, 5 |
749 |
|
AK081448.1 |
- |
4, 5 |
1019 |
|
AK081937.1 |
- |
4, 5 |
831 |
|
AK085099.1 |
- |
4, 5 |
1019 |
|
AK161786.1 |
- |
4, 5 |
912 |
|
BC057137.1 |
- |
4, 5 |
749 |
|
BC099954.1 |
- |
4, 5 |
845 |
30081872 |
BC141251.1 |
- |
4, 5 |
761 |
9055863 |
View Full Product Details