Gene Transcripts
- Gene Symbol
- Eml6
- Entrez Gene ID
- 237711
- Gene Name
- echinoderm microtubule associated protein like 6
- Gene Aliases
- 2900083P10Rik, C230094A16Rik, EMAP-6
- Chromosome Location
- Chr. 11: 29742586 - 30026034 on Build GRCm38
- UniGene ID
- Mm.445271
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146016.2 |
NP_666128.2
|
34, 35 |
5530 |
|
GenBank mRNA |
BC024726.1 |
- |
6, 7 |
965 |
|
View Full Product Details