Gene Transcripts

Gene Symbol
Mtr
Entrez Gene ID
238505
Gene Name
5-methyltetrahydrofolate-homocysteine methyltransferase
Gene Aliases
AI894170, D830038K18Rik, MS
Chromosome Location
Chr. 13: 12182720 - 12258195 on Build GRCm38
UniGene ID
Mm.40335
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081128.3 NP_001074597.1 20 2334
GenBank mRNA AK142258.1 - 20 2317
BC145683.1 - 20 2221 40130986
BC145685.1 - 20 2221 40130988

View Full Product Details