Gene Transcripts

Gene Symbol
Rimbp3
Entrez Gene ID
239731
Gene Name
RIMS binding protein 3
Gene Aliases
Gm1759, Gm603, RIM-BP3, mKIAA1666
Chromosome Location
Chr. 16: 17208135 - 17213982 on Build GRCm38
UniGene ID
Mm.86671
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033338.3 NP_001028510.2 1 3045
GenBank mRNA AK133200.1 - 1 2196
AK220410.1 - 1 1065
BC141386.1 - 1 2189 9055999

View Full Product Details