Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011909.2 | NP_036039.2 | 8 | 1008 | |
GenBank mRNA | AF069502.1 | - | 8 | 969 | |
AK149772.1 | - | 8 | 1009 | ||
AK151011.1 | - | 8 | 1009 | ||
AK151260.1 | - | 8 | 1009 | ||
AK151722.1 | - | 8 | 1009 | ||
AK152034.1 | - | 8 | 974 | ||
AK152574.1 | - | 8 | 974 | ||
AK153312.1 | - | 8 | 978 | ||
BC138562.1 | - | 8 | 890 | 8861582 | |
BC138577.1 | - | 8 | 890 | 8861597 | |
CT010228.1 | - | 7 | 744 |