Gene Transcripts

Gene Symbol
Atp8b4
Entrez Gene ID
241633
Gene Name
ATPase, class I, type 8B, member 4
Gene Aliases
A530043E15, Im
Chromosome Location
Chr. 2: 126320973 - 126500669 on Build GRCm38
UniGene ID
Mm.222235
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080944.3 NP_001074413.2 28 3662
GenBank mRNA AK155448.1 - 28 3672
AK169967.1 - 28 3690
BC130260.1 - 10 1636
BC152337.1 - 10 1636

View Full Product Details