Gene Transcripts

Gene Symbol
Fermt1
Entrez Gene ID
241639
Gene Name
fermitin family homolog 1 (Drosophila)
Gene Aliases
5830467P10Rik, Kindlin-1
Chromosome Location
Chr. 2: 132904177 - 132946036 on Build GRCm38
UniGene ID
Mm.209784
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198029.2 NP_932146.2 12 2246
GenBank mRNA AF073489.1 - 9 1032
AK030947.1 - 11 2436
AK050804.1 - 11 1394
BC029093.1 - 3 365

View Full Product Details