Gene Transcripts

Gene Symbol
Rxfp4
Entrez Gene ID
242093
Gene Name
relaxin family peptide receptor 4
Gene Aliases
GPCR142, Gpr100, Rln3r2, SALPR
Chromosome Location
Chr. 3: 88651898 - 88653142 on Build GRCm38
UniGene ID
Mm.269130
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181817.1 NP_861538.1 1 657
GenBank mRNA AY288422.1 - 1 657
AY633765.1 - 1 657
BC107269.1 - 1 657 40052601
BC107270.1 - 1 656

View Full Product Details