Gene Transcripts

Gene Symbol
Rsbn1l
Entrez Gene ID
242860
Gene Name
round spermatid basic protein 1-like
Gene Aliases
8430412F05Rik, AI447441, C330002G24Rik
Chromosome Location
Chr. 5: 20893024 - 20953441 on Build GRCm38
UniGene ID
Mm.254286
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080977.1 NP_001074446.1 3 1058
GenBank mRNA AK033374.1 - 3 738

View Full Product Details