Gene Transcripts

Gene Symbol
Cpz
Entrez Gene ID
242939
Gene Name
carboxypeptidase Z
Gene Aliases
AV136066
Chromosome Location
Chr. 5: 35502218 - 35525626 on Build GRCm38
UniGene ID
Mm.108557
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153107.2 NP_694747.2 1, 2 202
GenBank mRNA AF356844.1 - 1, 2 111
CJ067998.1 - 1, 2 274

View Full Product Details