Gene Transcripts

Gene Symbol
Nars2
Entrez Gene ID
244141
Gene Name
asparaginyl-tRNA synthetase 2 (mitochondrial)(putative)
Gene Aliases
AI875199
Chromosome Location
Chr. 7: 96951510 - 97064758 on Build GRCm38
UniGene ID
Mm.274864
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153591.4 NP_705819.3 14 1618
GenBank mRNA AK046799.1 - 14 1619
AK046801.1 - 14 1619
AK047091.1 - 14 1563
AK050718.1 - 14 1557
AK138260.1 - 14 1598
AK164179.1 - 14 1587
BC030041.1 - 10 822 1226297

View Full Product Details